Merck
  • Cytisine induces autonomic cardiovascular responses via activations of different nicotinic receptors.
Cytisine induces autonomic cardiovascular responses via activations of different nicotinic receptors.
Autonomic neuroscience : basic & clinical (2009-11-06)
Yi-Fan Li, Carly Lacroix, Jessica Freeling
ABSTRACT
Nicotinic cholinergic receptors mediate autonomic transmission at ganglia. However, whether different subtypes of nicotinic cholinergic receptors expressed in autonomic ganglia elicit distinct roles in mediating sympathetic and parasympathetic regulations remain to be defined. In this study, we observed that different subtypes of nicotinic receptors were responsible for the sympathetic and parasympathetic cardiovascular responses. In urethane anesthetized mice, intravenous injection with cytisine, a non-selective nicotinic agonist, induced a brief but pronounced decrease in heart rate, followed by increases in heart rate and arterial blood pressure. The bradycardic response was blocked by atropine, and the pressor response was blocked by prazosin, confirming that these responses were parasympathetic and sympathetic activities, respectively. Hexamethonium, a ganglionic blocker, blocked both sympathetic and parasympathetic responses. Pretreatment with methyllycaconitine citrate, a selective alpha7 nicotinic receptor antagonist, significantly attenuated cytisine-induced sympathetic response with little effect on the parasympathetic response. In contrast, pretreatment with dihydro-beta-erythroidine hydrobromide, a selective alpha4beta2 nicotinic receptor antagonist, blocked cytisine-induced parasympathetic response but not the sympathetic response. Pretreatment with dihydro-beta-erythroidine hydrobromide also blocked baroreflex associated parasympathetic bradycardic response. Moreover, treatment with nicotine induced a bradycardic response without a significant pressor response, which was also attenuated by dihydro-beta-erythroidine hydrobromide. Collectively, these data suggest that different nicotinic receptors play distinct roles in sympathetic and parasympathetic ganglia. Specifically, activations of alpha7 and alpha4beta2 nicotinic receptors are involved in cytisine-induced cardiovascular sympathetic and parasympathetic responses, respectively.