Merck
  • Dissection and Isolation of Murine Glia from Multiple Central Nervous System Regions.
Dissection and Isolation of Murine Glia from Multiple Central Nervous System Regions.
Journal of visualized experiments : JoVE (2020-06-23)
Maksim Sinyuk, Jessica L Williams
ABSTRACT
The methods presented here demonstrate laboratory procedures for the dissection of four different regions of the central nervous system (CNS) from murine neonates for the isolation of glial subpopulations. The purpose of the procedure is to dissociate microglia, oligodendrocyte progenitor cells (OPCs), and astrocytes from cortical, cerebellar, brainstem, and spinal cord tissue to facilitate further in vitro analysis. The CNS region isolation procedures allow for the determination of regional heterogeneity among glia in multiple cell culture systems. Rapid CNS region isolation is performed, followed by the mechanical removal of meninges to prevent meningeal cell contamination of glia. This protocol combines gentle tissue dissociation and plating on a specified matrix designed to preserve cell integrity and adherence. Isolating mixed glia from multiple CNS regions provides a comprehensive analysis of potentially heterogenous glia while maximizing the use of individual experimental animals. Additionally, following dissociation of regional tissue, mixed glia are further divided into multiple cell types including microglia, OPCs, and astrocytes for use in either single cell type, cell culture plate inserts, or co-culture systems. Overall, the demonstrated techniques provide a comprehensive protocol of broad applicability for careful dissection of four individual CNS regions from murine neonates and includes methods for the isolation of three individual glia cell types to examine regional heterogeneity in any number of in vitro cell culture systems or assays.