Merck
  • Human telomerase reverse transcriptase is a novel target of Hippo-YAP pathway.
Human telomerase reverse transcriptase is a novel target of Hippo-YAP pathway.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology (2020-01-18)
Qian Zhang, Ning Liu, Junjie Bai, Qi Zhou, Jian Mao, Lu Xu, Jiang Liu, Haibin Wei, Chengcheng Ren, Xiaoying Wu, Miao Wang, Bin Zhao, Yu-Sheng Cong
ABSTRACT
Telomerase plays a pivotal role in tumorigenesis by maintaining telomere homeostasis, a hallmark of cancer. However, the mechanisms by which telomerase is reactivated or upregulated during tumorigenesis remain incompletely understood. Here, we report that the Hippo pathway effector Yes-associated protein (YAP) regulates the expression of human telomerase reverse transcriptase (hTERT). Ectopic expression or physiological activation of YAP increases hTERT expression, whereas knockdown of YAP decreases the expression of hTERT. YAP binds to the hTERT promoter through interaction with the TEA domain family transcription factors and activates hTERT transcription. Furthermore, sustained YAP hyperactivation promotes telomerase activity and extends telomere length, with increased hTERT expression. In addition, we show that hTERT expression is positively correlated with YAP activation in human liver cancer tissues. Together, our results demonstrate that YAP promotes hTERT expression, which could contribute to tumor progression.