Merck
  • Combinatorial synthesis of anilinoanthraquinone derivatives and evaluation as non-nucleotide-derived P2Y2 receptor antagonists.
Combinatorial synthesis of anilinoanthraquinone derivatives and evaluation as non-nucleotide-derived P2Y2 receptor antagonists.
Bioorganic & medicinal chemistry letters (2007-11-17)
Stefanie Weyler, Younis Baqi, Petra Hillmann, Marko Kaulich, Andrea M Hunder, Ingrid A Müller, Christa E Müller
ABSTRACT
A library of anilinoanthraquinone derivatives was synthesized by parallel Ullmann coupling reaction of bromaminic acid with aniline derivatives in solution using a compact parallel synthesizer. The products were purified by HPLC and evaluated as antagonists at mouse and human P2Y2 receptors. 4-Phenylamino-substituted 1-amino-2-sulfoanthraquinones, for example, 1-amino-4-(2-methoxyphenyl)-2-sulfoanthraquinone (PSB-716), were potent P2Y2 antagonists with IC50 values in the low micromolar range.