- Bioactive secondary metabolites from Boesenbergia pandurata of Myanmar and their preferential cytotoxicity against human pancreatic cancer PANC-1 cell line in nutrient-deprived medium.
Bioactive secondary metabolites from Boesenbergia pandurata of Myanmar and their preferential cytotoxicity against human pancreatic cancer PANC-1 cell line in nutrient-deprived medium.
The chloroform extract of rhizomes of Boesenbergia pandurata demonstrated marked preferential cytotoxicity against human pancreatic PANC-1 cancer cells in nutrient-deprived medium. Bioactivity-directed investigation of this extract yielded four new secondary metabolites, geranyl-2,4-dihydroxy-6-phenethylbenzoate ( 1), 2',4'-dihydroxy-3'-(1''-geranyl)-6'-methoxychalcone ( 2), (1' R,2' S,6' R)-2-hydroxyisopanduratin A ( 3), and (2 R)-8-geranylpinostrobin ( 4), and twenty known compounds ( 5- 24). Among the known compounds, (2 S)-6-geranylpinostrobin ( 5), (+/-)-6-methoxypanduratin A ( 6), and (2 S)-7,8-dihydro-5-hydroxy-2-methyl-2-(4''-methyl-3''-pentenyl)-8-phenyl-2 H,6 H-benzo[1,2- b:5,4- b']dipyran-6-one ( 7) were isolated for the first time from a natural source. The structures of these compounds were elucidated using extensive spectroscopic techniques including CD measurements. All the isolated compounds showed varying degrees of in vitro preferential cytotoxicity against PANC-1 cells. Nicolaioidesin B ( 11) and panduratin A ( 17) were most potent, each showing a PC 100 at 2.5 microM.