Merck
  • Synthesis and characterization of cobalt(II), nickel(II), copper(II) and zinc(II) complexes with Schiff base derived from 4-amino-3-mercapto-6-methyl-5-oxo-1,2,4-triazine.
Synthesis and characterization of cobalt(II), nickel(II), copper(II) and zinc(II) complexes with Schiff base derived from 4-amino-3-mercapto-6-methyl-5-oxo-1,2,4-triazine.
European journal of medicinal chemistry (2007-01-17)
Kiran Singh, Manjeet Singh Barwa, Parikshit Tyagi
ABSTRACT
A few (1:1) and (1:2) metal complexes of cobalt(II), nickel(II), copper(II) and zinc(II) have been isolated with ligand derived from the condensation of 4-amino-3-mercapto-6-methyl-5-oxo-1,2,4-triazine with 2-acetylpyridine (L(1)) and characterized by elemental analysis, conductivity measurements, infrared, electronic, (1)H NMR spectral data, magnetic and thermogravimetric analyses. Due to insolubility in water and most of the common organic solvents and infusibility at higher temperatures, all the complexes are thought to be polymeric in nature. A square-planar geometry was suggested for copper(II) and octahedral proposed for cobalt(II), nickel(II) and zinc(II). Some of the chemically synthesized compounds have been screened in vitro against the three Gram-positive (Staphylococcus aureus, Staphylococcus epidermidis and Bacillus subtilis) and two Gram-negative (Salmonella typhi and Escherichia coli) organisms. It is observed that the coordination of metal ion has pronounced effect on the microbial activities of the ligand. The metal complexes have higher antimicrobial effect than the free ligands.