Merck
  • Glutamate potentiates the toxicity of mutant Cu/Zn-superoxide dismutase in motor neurons by postsynaptic calcium-dependent mechanisms.
Glutamate potentiates the toxicity of mutant Cu/Zn-superoxide dismutase in motor neurons by postsynaptic calcium-dependent mechanisms.
The Journal of neuroscience : the official journal of the Society for Neuroscience (1998-11-21)
J Roy, S Minotti, L Dong, D A Figlewicz, H D Durham
ABSTRACT
Mutations in the Cu/Zn-superoxide dismutase (SOD-1) gene are responsible for a subset of familial cases of amyotrophic lateral sclerosis. Using a primary culture model, we have demonstrated that normally nontoxic glutamatergic input, particularly via calcium-permeable AMPA/kainate receptors, is a major factor in the vulnerability of motor neurons to the toxicity of SOD-1 mutants. Wild-type and mutant (G41R, G93A, or N139K) human SOD-1 were expressed in motor neurons of dissociated cultures of murine spinal cord by intranuclear microinjection of plasmid expression vector. Both a general antagonist of AMPA/kainate receptors (CNQX) and a specific antagonist of calcium-permeable AMPA receptors (joro spider toxin) reduced formation of SOD-1 proteinaceous aggregates and prevented death of motor neurons expressing SOD-1 mutants. Partial protection was obtained by treatment with nifedipine, implicating Ca2+ entry through voltage-gated calcium channels as well as glutamate receptors in potentiating the toxicity of mutant SOD-1 in motor neurons. Dramatic neuroprotection was obtained by coexpressing the calcium-binding protein calbindin-D28k but not by increasing intracellular glutathione levels or treatment with the free radical spin trap agent, N-tert-butyl-alpha-phenylnitrone. Thus, generalized oxidative stress could have contributed in only a minor way to death of motor neurons expressing the mutant SOD-1. These studies demonstrated that the toxicity of these mutants is calcium-dependent and provide direct evidence that calcium entry during neurotransmission, coupled with deficiency of cytosolic calcium-binding proteins, is a major factor in the preferential vulnerability of motor neurons to disease.