- Anthraquinone based polymer as high performance cathode material for rechargeable lithium batteries.
Anthraquinone based polymer as high performance cathode material for rechargeable lithium batteries.
Chemical communications (Cambridge, England) (2009-01-13)
Zhiping Song, Hui Zhan, Yunhong Zhou
PMID19137181
ABSTRACT
Poly(anthraquinonyl sulfide) is synthesized and investigated as a novel organic cathode material for rechargeable lithium batteries, which shows excellent reversibility and cyclabilty, and gives important insights into developing a new generation of organic cathode materials with higher performance.