Merck
  • Low-Concentration Essential Amino Acids in PZM-3 Improve the Developmental Competence of Porcine Embryos Produced by Handmade Cloning.
Low-Concentration Essential Amino Acids in PZM-3 Improve the Developmental Competence of Porcine Embryos Produced by Handmade Cloning.
Cellular reprogramming (2020-11-13)
Tianbin Liu, Jie Li, Hongwei Dou, Xi Xiang, Wenbin Chen, Tingting Zhang, Lin Li, Xingju Zhang, Xuan Dong, Lei Chen, Xuyi Lin, Jing Li, Hai-Xi Sun, Ying Gu, Lin Lin
ABSTRACT
Essential amino acids (EAA) of inappropriate concentration have been reported to compromise the development of embryo. This study aimed to investigate the effect of EAA on the developmental competence of porcine embryos produced by either handmade cloning (HMC) or parthenogenetic activation (PA). In experiment 1, we examined the in vitro developmental competence of PA embryos after culture in PZM-3 containing different concentrations (v/v) of EAA (0%, 1%, and 2%). The results indicated that reducing the concentration of EAA from 2% to 1% significantly improved the blastocyst formation (36% vs. 54%), while 0% would compromise the blastocyst formation rate (54% vs. 38%). In experiment 2, we further investigated the effect of EAA concentration (1% and 2%) on the in vitro developmental competence and gene expression of HMC embryos. Blastocyst rate significantly increased by reducing concentration of EAA (41% vs. 53%) and those genes upregulated were enriched in oxidative phosphorylation, PPAR signaling pathway, and metabolism-related pathways. In experiment 3, the in vivo developmental competence of HMC embryos cultured in the medium supplemented with 1% EAA was examined. Embryos derived from both non-gene-modified fetal fibroblasts (FFs) and gene-modified fetal fibroblasts (GMFFs) were transferred to recipients. The pregnancy rates were 83% and 78% separately. Out of the pregnancies, 5 (FFs) and 6 (GMFFs) were successfully developed to term. Our study indicates that supplementing EAA to embryo culture medium at a concentration of 1% can improve the in vitro developmental competence of porcine HMC embryos and the blastocyst obtained can successfully develop to term, which could be beneficial for the production of gene-modified piglets.