Merck
  • Elevation of JAML Promotes Diabetic Kidney Disease by Modulating Podocyte Lipid Metabolism.
Elevation of JAML Promotes Diabetic Kidney Disease by Modulating Podocyte Lipid Metabolism.
Cell metabolism (2020-11-14)
Yi Fu, Yu Sun, Mei Wang, Yunfeng Hou, Wei Huang, Di Zhou, Ziying Wang, Shuting Yang, Wei Tang, Junhui Zhen, Yujia Li, Xiaojie Wang, Min Liu, Yan Zhang, Baobao Wang, Guangyi Liu, Xiao Yu, Jinpeng Sun, Chun Zhang, Fan Yi
ABSTRACT
Lipid accumulation in podocytes is a major determinant of diabetic kidney disease (DKD) and identification of potential therapeutic targets by mediating podocyte lipid metabolism has clinical importance. This study was to elucidate the role of JAML (junctional adhesion molecule-like protein) in the pathogenesis of DKD. We first confirmed the expression of JAML in podocytes and found that podocyte-specific deletion of Jaml ameliorated podocyte injury and proteinuria in two different models of diabetic mice. We further demonstrated a novel role of JAML in regulating podocyte lipid metabolism through SIRT1-mediated SREBP1 signaling. Similar results were also found in mice with adriamycin-induced nephropathy. Importantly, we observed a higher expression of JAML in glomeruli from subjects with DKD and other types of proteinuric kidney diseases, and the level of JAML was correlated with lipid accumulation and glomerular filtration rate, suggesting that JAML may be an attractive therapeutic target for proteinuric kidney disease.