Merck
  • Ligand-specific alphabeta and gammadelta T cell responses in childhood tuberculosis.
Ligand-specific alphabeta and gammadelta T cell responses in childhood tuberculosis.
The Journal of infectious diseases (1999-12-23)
F Dieli, G Sireci, C Di Sano, A Romano, L Titone, P Di Carlo, J Ivanyi, J J Fourniè, A Salerno
ABSTRACT
The alphabeta and gammadelta T cell responses were analyzed in the peripheral blood of children affected by active tuberculosis (TB) and in healthy children who tested positive (PPD+) or negative (PPD-) for purified protein derivative. PPD+ healthy and diseased children responded equally well to PPD in vitro. In contrast, only 18% of PPD+ TB patients responded to peptide p38G derived from the 38-kDa protein of Mycobacterium tuberculosis. Analysis of the whole gammadelta T cell population and of its Vgamma9/Vdelta2 subset showed similar frequencies in PPD+ children with TB and in healthy PPD+ and PPD- children. Vgamma9/Vdelta2 cells from children with TB responded to 5 different phosphoantigens similarly to those from healthy PPD+ children, but healthy PPD- children responded very poorly. Chemotherapy had contrasting effects on the tested lymphocyte population, represented by increase of alphabeta and decline of Vgamma9/Vdelta2 T cell responses. T cell responses in childhood TB may be similar to those in adult TB.