Merck
  • Protein Gq modulates termination of phototransduction and prevents retinal degeneration.
Protein Gq modulates termination of phototransduction and prevents retinal degeneration.
The Journal of biological chemistry (2012-03-06)
Wen Hu, Didi Wan, Xiaoming Yu, Jinguo Cao, Peiyi Guo, Hong-Sheng Li, Junhai Han
ABSTRACT
Appropriate termination of the phototransduction cascade is critical for photoreceptors to achieve high temporal resolution and to prevent excessive Ca(2+)-induced cell toxicity. Using a genetic screen to identify defective photoresponse mutants in Drosophila, we isolated and identified a novel Gα(q) mutant allele, which has defects in both activation and deactivation. We revealed that G(q) modulates the termination of the light response and that metarhodopsin/G(q) interaction affects subsequent arrestin-rhodopsin (Arr2-Rh1) binding, which mediates the deactivation of metarhodopsin. We further showed that the Gα(q) mutant undergoes light-dependent retinal degeneration, which is due to the slow accumulation of stable Arr2-Rh1 complexes. Our study revealed the roles of G(q) in mediating photoresponse termination and in preventing retinal degeneration. This pathway may represent a general rapid feedback regulation of G protein-coupled receptor signaling.