Merck
Dopamine receptors in the human dura mater.
Neurochemical research (2004-07-21)
Carlo Cavallotti, Daniela Cavallotti, Gianfranco Tonnarini, Francesca Maria Tranquilli Leali
ABSTRACT
Dopamine receptors (Dar) were studied as a component of the nervous dopaminergic system in the human dura mater. Dar were stained in several dural zones (vascular, perivascular, intervascular) in different regions (basal, calvarial, tentorial, occipital, frontal, parietal, temporal) of the cranial meninges. Specimens of human dura mater were harvested from autopsies of 10 elderly male subjects (age range, 60-75 years). Dar were labeled with specific (H3) markers, studied with radiobinding techniques (including liquid scintillation), stained for light microscope autoradiography, and measured by means of quantitative analysis of images. All results were evaluated with statistical analysis to identify significant results. More dural Dar were found in the basal region than in the calvarial one. Moreover, Dar are more abundant in the vascular and perivascular dural zone than in the intervascular one. The vascular distribution of Dar seemed to indicate that Dar play a role in the control of meningeal blood vessels. The location and distribution of D1 and D2 receptors in the human cranial dura mater confirmed the presence of a dopaminergic system, which could play an important role in controlling blood flow and/or other functions of meningeal membranes.
Sigma-Aldrich
Anti-Dopamine D1B Receptor Antibody, clone SG4-D1B, clone SG4-D1B, Chemicon®, from mouse
Sigma-Aldrich
Anti-Dopamine D1A Receptor Antibody, clone SG2-D1a, clone SG2-D1a, Chemicon®, from mouse