Merck
  • Development, optimization and validation of an absolute specific assay for active myeloperoxidase (MPO) and its application in a clinical context: role of MPO specific activity in coronary artery disease.
Development, optimization and validation of an absolute specific assay for active myeloperoxidase (MPO) and its application in a clinical context: role of MPO specific activity in coronary artery disease.
Clinical chemistry and laboratory medicine (2020-02-08)
Alessandro Trentini, Valentina Rosta, Savino Spadaro, Tiziana Bellini, Paola Rizzo, Francesco Vieceli Dalla Sega, Angelina Passaro, Giovanni Zuliani, Valentina Gentili, Gianluca Campo, Carlo Cervellati
ABSTRACT
Background Myeloperoxidase (MPO) is an enzyme with a recognized prognostic role in coronary artery disease (CAD), which is also emerging as a promising biomarker for cardiac risk stratification. However, the lack of a consensus method for its quantification has hindered its implementation in clinical practice. The aim of our work was to optimize an absolute sensitive assay for active MPO without external standards, to validate the method in the clinical context of CAD patients, and to estimate the enzyme specific activity. Methods In order to determine the MPO concentration using fluorescence readings, this ELISA assay exploits the activity of the enzyme recognized by specific antibodies. The assay was validated in a small cohort of patients that included: healthy subjects (n=60); patients with acute myocardial infarction (AMI, n=25); patients with stable CAD (SCAD, n=25) and a concomitant chronic obstructive pulmonary disease (COPD). Then, total MPO concentration and specific activity (activity/total MPO) were determined. Results The assay showed an intra- and inter-assay coefficient of variation of 5.8% and 10.4%, respectively, with a limit of detection (LoD) of 0.074 μU. Both AMI and SCAD patients had higher active and total MPO than controls (p<0.0001 and p<0.01, respectively). The specific activity of MPO was higher in SCAD patients compared to both controls and AMI (p<0.0001). Conclusions The study presents a robust and sensitive method for assaying MPO activity in biological fluids with low variability. Moreover, the determination of the specific activity could provide novel insight into the role of MPO in cardiovascular diseases (CVDs).