Merck
  • Synthesis and efficacy of 1-[bis(4-fluorophenyl)-methyl]piperazine derivatives for acetylcholinesterase inhibition, as a stimulant of central cholinergic neurotransmission in Alzheimer's disease.
Synthesis and efficacy of 1-[bis(4-fluorophenyl)-methyl]piperazine derivatives for acetylcholinesterase inhibition, as a stimulant of central cholinergic neurotransmission in Alzheimer's disease.
Bioorganic & medicinal chemistry letters (2006-06-01)
C T Sadashiva, J N Narendra Sharath Chandra, K C Ponnappa, T Veerabasappa Gowda, Kanchugarakoppal S Rangappa
ABSTRACT
The cholinergic hypothesis of Alzheimer's disease (AD) has spurred the development of numerous structural classes of compounds with different pharmacological profile aimed at increasing central cholinergic neurotransmission. Thus proving a symptomatic treatment for this disease are cholinomimetics with the pharmacological profile of acetyl cholinesterase (AchE) inhibitors. The novel bioactive 1-[bis(4-fluorophenyl)-methyl]piperazine derivatives were synthesized under mild conditions using different aryl/alkyl halides and heterocyclic alkyl halides with 1-[bis(4-fluorophenyl)-methyl]piperazine in the presence of powdered potassium carbonate in N,N-dimethylformamide. All the synthesized compounds were characterized by spectroscopic techniques, elemental analysis and were screened for their efficacy as AchE inhibitor. Some derivatives in this class showed good inhibition against AchE as compared to neostigmine as standard.