- Cytotoxic principles from the formosan milkweed, Asclepias curassavica.
Cytotoxic principles from the formosan milkweed, Asclepias curassavica.
Journal of natural products (2005-10-29)
Michael C Roy, Fang-Rong Chang, Hsiao-Chu Huang, Michael Y-N Chiang, Yang-Chang Wu
PMID16252914
ABSTRACT
A series of cardenolides and related compounds have been isolated from the aerial parts and roots of the ornamental milkweed, Asclepias curassavica. Their structures were determined by spectroscopic and chemical methods. Among them, three derivatives of calactinic acid methyl ester (13-15), 19-nor-16 alpha-acetoxy-10 beta-hydroxyasclepin (16), 20 beta,21-dihydroxypregna-4,6-dien-3-one (19), and 3,4-seco-urs-20(30)-en-3-oic acid (22) are new compounds. The relative configuration of calactinic acid methyl ester (12) has been confirmed by X-ray diffraction analysis on its derivative 13. Most of the cardenolides obtained showed pronounced cytotoxicity against four cancer cell lines (IC(50) 0.01 to 2.0 microg/mL).