Merck
  • Synthesis and biological properties of new stilbene derivatives of resveratrol as new selective aryl hydrocarbon modulators.
Synthesis and biological properties of new stilbene derivatives of resveratrol as new selective aryl hydrocarbon modulators.
Journal of medicinal chemistry (2005-01-07)
Philippe de Medina, Robert Casper, Jean-François Savouret, Marc Poirot
ABSTRACT
We developed new stilbene derivatives of resveratrol (E)-1-(4'-hydroxyphenyl)-2-(3,5-dihydroxyphenyl)ethene) selective for AhR and devoid of affinity for ER. Among the 24 stilbenes synthesized, all display a higher affinity than resveratrol for AhR. (E)-1-(4'-Trifluoromethylphenyl)-2-(3,5-ditrifluoromethylphenyl)ethene (4e), (E)-1-(4'-methoxyphenyl)-2-(3,5-dichlorophenyl)ethene (4j), and (E)-1-(4'-chlorophenyl)-2-(3,5-dichlorophenyl)ethene (4b) are selective, high-affinity AhR antagonists with, respective, K(i)s of 2.1, 1.4, and 1.2 nM. (E)-1-(4'-Trifluoromethylphenyl)-2-(3,5-dichlorophenyl)ethene (4i) displays a K(i) of 0.2 nM and is a selective and high-affinity agonist on AhR.
Sigma-Aldrich
Resveratrol, ≥99% (HPLC)