Merck
  • Disulfide polymer grafted polypropylene/polyethylene filter media for selective cadmium removal.
Disulfide polymer grafted polypropylene/polyethylene filter media for selective cadmium removal.
Journal of hazardous materials (2020-09-18)
Jeongmin Hong, Dongah Ko, Yuhoon Hwang
ABSTRACT
Heavy metal pollution caused by stormwater runoff has triggered a demand for effective heavy metal sorbents. Effective heavy metal removal using conventional stormwater runoff treatment processes that employ filtration mechanisms as primary removal mechanisms is difficult. Therefore, we attempt to improve cadmium removal performance by attaching disulfide polymer (DiS-COP) containing soft bases, thiols, onto the surface of polypropylene/polyethylene (PP/PE) fiber media, which is widely used for stormwater runoff treatment. Material characterization demonstrated that DiS-COP was successfully grafted and grown on the surface of PP/PE (Dis-PP/PE). The batch and continuous flow adsorption capacities of Dis-PP/PE were 81.1 mg/g and 2.33 mg/g, respectively, which is 40 times higher than those of pristine PP/PE. Applicability of DiS-PP/PE at pH 6-8 was demonstrated, and effects of calcium and humic acid on cadmium adsorption were investigated. Calcium marginally affected cadmium adsorption, which can be explained using the Hard and soft (Lewis) acids and bases theory (HSAB), but cadmium removal efficiency decreased owing to humic acid (HA)-Cd complex formation and agglomeration in the presence of organic material. In a breakthrough test, the adsorption column exhibited complete cadmium uptake over 24 h until it reached the breakthrough point. Therefore, heavy metal adsorption performance of PP/PE was successfully enhanced by grafting DiS-COP on its surface.