Merck
  • Optimization of Insecticidal Triterpene Derivatives by Biomimetic Oxidations with Hydrogen Peroxide and Iodosobenzene Catalyzed by MnIII and FeIII Porphyrin Complexes.
Optimization of Insecticidal Triterpene Derivatives by Biomimetic Oxidations with Hydrogen Peroxide and Iodosobenzene Catalyzed by MnIII and FeIII Porphyrin Complexes.
Chemistry & biodiversity (2020-07-10)
Noureddine Mazoir, Ahmed Benharref, Laura Vaca, Matías Reina, Azucena González-Coloma
ABSTRACT
Semisynthetic functionalized triterpenes (4α,14-dimethyl-5α,8α-8,9-epoxycholestan-3β-yl acetate; 4α,14-dimethyl-5α-cholest-8-ene-3,7,11-trione; 4α,14-dimethyl-5α-cholesta-7,9(11)-dien-3-one and 4α,14-dimethyl-5α-cholest-8-en-3β-yl acetate), previously prepared from 31-norlanostenol, a natural insecticide isolated from the latex of Euphorbia officinarum, have been subjected to oxidation with hydrogen peroxide (H2 O2 ) and iodosobenzene (PhIO) catalyzed by porphyrin complexes (cytochrome P-450 models) in order to obtain optimized derivatives with high regioselectivity. The main transformations were epoxidation of the double bonds and hydroxylations of non-activated C-H groups and the reaction products were 25-hydroxy-4α,14-dimethyl-5α-cholesta-7,9(11)-dien-3β-yl acetate (59 %), 25-hydroxy-4α,14-dimethyl-5α-cholest-8-ene-3,7,11-trione (60 %), 4α,14-dimethyl-5α,7β-7,8-epoxycholest-9(11)-en-3-one (22 %), 8-hydroxy-4α,14-dimethyl-5α-cholest-9(11)-ene-3,7-dione (16 %), 12α-hydroxy-4α,14-dimethyl-5α,7β-7,8-epoxycholest-9(11)-en-3-one (16 %), and 4α,14-dimethyl-5α,8α-8,9-epoxycholestan-3β-yl acetate (26 %), respectively. We also investigated the insect (Myzus persicae, Rhopalosiphum padi and Spodoptera littoralis) antifeedant and postingestive effects of these terpenoid derivatives. None of the compounds tested had significant antifeedant effects, however, all were more effective postingestive toxicants on S. littoralis larvae than the natural compound 31-norlanostenol, with 4α,14-dimethyl-5α,8α-8,9-epoxycholestan-3β-yl acetate being the most active. The study of their structure-activity relationships points out at the importance of C3 and C7 substituents.