Merck
  • MicroRNA-301a promotes pancreatic cancer invasion and metastasis through the JAK/STAT3 signaling pathway by targeting SOCS5.
MicroRNA-301a promotes pancreatic cancer invasion and metastasis through the JAK/STAT3 signaling pathway by targeting SOCS5.
Carcinogenesis (2019-06-25)
Hui Hu, Qin Zhang, Weiqun Chen, Tangwei Wu, Shuiyi Liu, Xiaoyi Li, Bo Luo, Tianzhu Zhang, Ge Yan, Hongda Lu, Zhongxin Lu
ABSTRACT
Pancreatic cancer is one of the most lethal digestive malignant tumors. We had previously found that microRNA-301a (miR-301a) is a oncogenic microRNA whose recognized conduce to nuclear factor-kappa B (NF-κB) activation in pancreatic cancer, yet the underlying mechanisms of miR-301a in promoting pancreatic cancer invasion and migration is obscure. In this work we found that high expression of miR-301a in human pancreatic cancer patients is related to poor survival. Overexpression of miR-301a enhances pancreatic cancer cell invasion, angiogenesis and migration, whereas inhibition of miR-301a suppresses pancreatic cancer cell invasion and reduces orthotopic pancreatic tumor growth and metastasis. Furthermore, suppressor of cytokine signaling 5 (SOCS5) is identified as a target gene of miR-301a. We found that miR-301a suppressed the expression of SOCS5 leads to janus kinase/signal transducer and activator of transcription 3 (JAK/STAT3) activation and is related to poor overall survival of pancreatic cancer patients. Taken together, our data show for the first time that the feedback loop between miR-301a and JAK/STAT3 pathway may play a significant role in pancreatic cancer invasion and metastasis. Targeting the loop may prove beneficial to prevent metastasis and provide a more effective therapeutic strategy for pancreatic cancer.