Merck
  • Curcumin may reverse 5-fluorouracil resistance on colonic cancer cells by regulating TET1-NKD-Wnt signal pathway to inhibit the EMT progress.
Curcumin may reverse 5-fluorouracil resistance on colonic cancer cells by regulating TET1-NKD-Wnt signal pathway to inhibit the EMT progress.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie (2020-09-06)
Yi Lu, Runzhe Zhang, Xinjie Zhang, Bo Zhang, Qinghua Yao
ABSTRACT
Colorectal cancer is a kind of gastrointestinal tumor with rising morbidity and mortality. 5-fluorouracil is one of the most effective chemotherapy drugs for the treatment of CRC. However, clinical data reported dramatic resistance on the treatment for CRC with 5-fluorouracil. Present study aims to explore the anti-resistant effect of curcumin and its mechanism. MTT assay was used to evaluate the proliferation of rHCT-116 cells. Flow cytometry was used to determine the apoptosis and cell cycle of rHCT-116 cells. Western Blot was performed to detect the expression level of TET1, NKD2, E-cadherin, Vimentin, β-catenin, TCF4 and Axin in transfected rHCT-116 cells. 5-fluorouracil resistant HCT-116 cells were successfully established. Curcumin was found to be effective in the inhibition of proliferation, inducement of apoptosis and block of G0/G1 phase on 5-fluorouracil treated HCT-116 cells. The expression of TET1 and NKD2 was greatly inhibited by high dosage of curcumin. The WNT signal pathway and EMT progress were suppressed in rHCT-116 cells by high dosage of curcumin. The inhibitory effects of curcumin on WNT signal pathway and EMT progress were verified to be consistent with Pax-6, TET1 and NKD2. Curcumin might exert anti-resistant effect of 5-FU on HCT-116 cells by regulating the TET1-NKD2-WNT signal pathway to inhibit the EMT progress.