Merck
  • Investigation of Strand-Selective Interaction of SNA-Modified siRNA with AGO2-MID.
Investigation of Strand-Selective Interaction of SNA-Modified siRNA with AGO2-MID.
International journal of molecular sciences (2020-07-29)
Yukiko Kamiya, Yuuki Takeyama, Tomonari Mizuno, Fuminori Satoh, Hiroyuki Asanuma
ABSTRACT
Small interfering RNA (siRNA) has been recognized as a powerful gene-silencing tool. For therapeutic application, chemical modification is often required to improve the properties of siRNA, including its nuclease resistance, activity, off-target effects, and tissue distribution. Careful siRNA guide strand selection in the RNA-induced silencing complex (RISC) is important to increase the RNA interference (RNAi) activity as well as to reduce off-target effects. The passenger strand-mediated off-target activity was previously reduced and on-target activity was enhanced by substitution with acyclic artificial nucleic acid, namely serinol nucleic acid (SNA). In the present study, the reduction of off-target activity caused by the passenger strand was investigated by modifying siRNAs with SNA. The interactions of SNA-substituted mononucleotides, dinucleotides, and (2,2,6,6-tetramethylpiperidin-1-yl)oxyl (TEMPO)-labeled double-stranded RNA (dsRNA) with the MID domain of the Argonaute 2 (AGO2) protein, which plays a pivotal role in strand selection by accommodation of the 5'-terminus of siRNA, were comprehensively analyzed. The obtained nuclear magnetic resonance (NMR) data revealed that AGO2-MID selectively bound to the guide strand of siRNA due to the inhibitory effect of the SNA backbone located at the 5' end of the passenger strand.