Merck
  • Generation of an ALS human iPSC line KEIOi001-A from peripheral blood of a Charcot disease-affected patient carrying TARDBP p.N345K heterozygous SNP mutation.
Generation of an ALS human iPSC line KEIOi001-A from peripheral blood of a Charcot disease-affected patient carrying TARDBP p.N345K heterozygous SNP mutation.
Stem cell research (2020-07-14)
Nicolas Leventoux, Satoru Morimoto, Kenju Hara, Shiho Nakamura, Fumiko Ozawa, Shio Mitsuzawa, Tetsuya Akiyama, Ayumi Nishiyama, Naoki Suzuki, Hitoshi Warita, Masashi Aoki, Hideyuki Okano
ABSTRACT
Amyotrophic Lateral Sclerosis is the most common motor neuron degenerative disease in adults, and TARDBP gene mutations have been reported to be involved in the pathogenesis. We present here how we generated the human induced pluripotent stem cell (hiPSC) line KEIOi001-A/SM4-4-5 from the peripheral blood of a 63-year-old male patient presenting the c.1035C > G heterozygous SNP mutation in the TARDBP gene locus. The established hiPSC line does not express the exogenous reprogramming factors oriP nor EBNA1 and shows no karyotypic abnormalities, while it expresses pluripotent stem cell markers, presents the SNP mutation and is capable of three-germ layers differentiation in vitro.
Sigma-Aldrich
SIGMAFAST BCIP®/NBT, tablet