Merck
  • Beta-adrenergic receptor activation during distinct patterns of stimulation critically modulates the PKA-dependence of LTP in the mouse hippocampus.
Beta-adrenergic receptor activation during distinct patterns of stimulation critically modulates the PKA-dependence of LTP in the mouse hippocampus.
Learning & memory (Cold Spring Harbor, N.Y.) (2008-04-29)
Jennifer N Gelinas, Gustavo Tenorio, Neal Lemon, Ted Abel, Peter V Nguyen
ABSTRACT
Activation of beta-adrenergic receptors (beta-ARs) enhances hippocampal memory consolidation and long-term potentiation (LTP), a likely mechanism for memory storage. One signaling pathway linked to beta-AR activation is the cAMP-PKA pathway. PKA is critical for the consolidation of hippocampal long-term memory and for the expression of some forms of long-lasting hippocampal LTP. How does beta-AR activation affect the PKA-dependence, and persistence, of LTP elicited by distinct stimulation frequencies? Here, we use in vitro electrophysiology to show that patterns of stimulation determine the temporal phase of LTP affected by beta-AR activation. In addition, only specific patterns of stimulation recruit PKA-dependent LTP following beta-AR activation. Impairments of PKA-dependent LTP maintenance generated by pharmacologic or genetic deficiency of PKA activity are also abolished by concurrent activation of beta-ARs. Taken together, our data show that, depending on patterns of synaptic stimulation, activation of beta-ARs can gate the PKA-dependence and persistence of synaptic plasticity. We suggest that this may allow neuromodulatory receptors to fine-tune neural information processing to meet the demands imposed by numerous synaptic activity profiles. This is a form of "metaplasticity" that could control the efficacy of consolidation of hippocampal long-term memories.