Merck
MEK1/2 dual-specificity protein kinases: structure and regulation.
Biochemical and biophysical research communications (2011-12-20)
Robert Roskoski
ABSTRACT
MEK1 and MEK2 are related protein kinases that participate in the RAS-RAF-MEK-ERK signal transduction cascade. This cascade participates in the regulation of a large variety of processes including apoptosis, cell cycle progression, cell migration, differentiation, metabolism, and proliferation. Moreover, oncogenic mutations in RAS or B-RAF are responsible for a large proportion of human cancers. MEK1 is activated by phosphorylation of S218 and S222 in its activation segment as catalyzed by RAF kinases in an intricate process that involves a KSR scaffold. Besides functioning as a scaffold, the kinase activity of KSR is also required for MEK activation. MEK1 regulation is unusual in that S212 phosphorylation in its activation segment is inhibitory. Moreover, active ERK catalyzes a feedback inhibitory phosphorylation of MEK1 T292 that serves to downregulate the pathway.