Merck
  • Genetic polymorphisms of human flavin-containing monooxygenase 3: implications for drug metabolism and clinical perspectives.
Genetic polymorphisms of human flavin-containing monooxygenase 3: implications for drug metabolism and clinical perspectives.
Pharmacogenomics (2007-06-15)
Irfan M Hisamuddin, Vincent W Yang
ABSTRACT
Flavin-containing monooxygenase 3 (FMO3) is a hepatic microsomal enzyme that oxidizes a host of drugs, xenobiotics and other chemicals. Numerous variants in the gene encoding FMO3 have been identified, some of which result in altered enzymatic activity and, consequently, altered substrate metabolism. Studies also implicate individual and ethnic differences in the frequency of FMO3 polymorphisms. In addition, new variants continue to be identified with potentially important clinical implications. For example, the role of FMO3 variants in the pathophysiology of gastrointestinal diseases is an evolving area of research. Two commonly occurring polymorphisms of FMO3, E158K and E308G, have been associated with a reduction in polyp burden in patients with familial adenomatous polyposis who were treated with sulindac sulfide, an FMO3 substrate. These findings suggest a potential role for prospective genotyping of common FMO3 polymorphisms in the treatment of disease states that involve the use of drugs metabolized by FMO3. This review summarizes the current state of research on the genetic polymorphisms of FMO3, with a focus on their clinical implications in gastrointestinal diseases.