Merck
  • DPP-4 inhibitor reduces striatal microglial deramification after sensorimotor cortex injury induced by external force impact.
DPP-4 inhibitor reduces striatal microglial deramification after sensorimotor cortex injury induced by external force impact.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology (2020-04-05)
Yu-Wen Hung, Yun Wang, Sheau-Ling Lee
ABSTRACT
Dipeptidyl peptidase-4 inhibitors (or gliptins), a class of antidiabetic drugs, have recently been shown to have protective actions in the central nervous system. Their cellular and molecular mechanisms responsible for these effects are largely unknown. In the present study, two structurally different gliptins, sitagliptin and vildagliptin, were examined for their therapeutic actions in a controlled cortical impact (CCI) model of moderate traumatic brain injury (TBI) in mice. Early post-CCI treatment with sitagliptin, but not vildagliptin, significantly reduced body asymmetry, locomotor hyperactivity, and brain lesion volume. Sitagliptin attenuated post-CCI microglial deramification in the ipsilateral dorsolateral (DL) striatum, while vildagliptin had no effect. Sitagliptin also reduced striatal expression of galectin-3 and monocyte chemoattractant protein 1(MCP-1), and increased the cortical and striatal levels of the anti-inflammatory cytokine IL-10 on the ipsilateral side. These data support a differential protective effect of sitagliptin against TBI, possibly mediated by an anti-inflammatory effect in striatum to preserve connective network. Both sitagliptin and vildagliptin produced similar increases of active glucagon-like peptide-1 (GLP-1) in blood and brain. Increasing active GLP-1 may not be the sole molecular mechanisms for the neurotherapeutic effect of sitagliptin in TBI.
Product Description
Millipore
2,3,5-Triphenyltetrazolium chloride, Color indicator. Supplement in culture media for the microbiological analysis of food and water samples