Merck
  • Diagnostic and prognostic values of PBMC proteins in amyotrophic lateral sclerosis.
Diagnostic and prognostic values of PBMC proteins in amyotrophic lateral sclerosis.
Neurobiology of disease (2020-02-23)
Silvia Luotti, Laura Pasetto, Luca Porcu, Valter Torri, Saioa R Elezgarai, Serena Pantalone, Melania Filareti, Massimo Corbo, Christian Lunetta, Gabriele Mora, Valentina Bonetto
ABSTRACT
Amyotrophic lateral sclerosis (ALS) is a fatal motor neuron disease for which there are no validated biomarkers. Previous exploratory studies have identified a panel of candidate protein biomarkers in peripheral blood mononuclear cells (PBMCs) that include peptidyl-prolyl cis-trans isomerase A (PPIA), heat shock cognate protein 71 kDa (HSC70), heterogeneous nuclear ribonucleoprotein A2/B1 (hnRNPA2B1) and TDP-43. It has also been found that PPIA plays a key role in the assembly and dynamics of ribonucleoprotein (RNP) complexes and interacts with TDP-43. Its absence accelerates disease progression in a SOD1 mouse model of ALS, and low levels of PPIA in PBMCs are associated with early-onset ALS. However, the diagnostic and prognostic values of PPIA and the other candidate protein biomarkers have not been established. We analyzed the PBMC proteins in a well-characterized cohort of ALS patients (n=93), healthy individuals (n=104) and disease controls (n=111). We used a highly controlled sample processing procedure that implies two-step differential detergent fractionation. We found that the levels of the selected PBMC proteins in the soluble and insoluble fraction, combined, have a high discriminatory power for distinguishing ALS from controls, with PPIA, hnRNPA2B1 and TDP-43 being the proteins most closely associated with ALS. We also found a shift toward increased protein partitioning in the insoluble fraction in ALS and this correlated with a worse disease phenotype. In particular, low PPIA soluble levels were associated with six months earlier death. In conclusion, PPIA is a disease modifier with prognostic potential. PBMC proteins indicative of alterations in protein and RNA homeostasis are promising biomarkers of ALS, for diagnosis, prognosis and patient stratification.
Product Description
Millipore
ReBlot Plus Kit