Merck
  • Postnatal distribution of ZnO nanoparticles to the breast milk through oral route and their risk assessment for breastfed rat offsprings.
Postnatal distribution of ZnO nanoparticles to the breast milk through oral route and their risk assessment for breastfed rat offsprings.
Human & experimental toxicology (2020-04-30)
A Hussain, S Kumar, G Kaul
ABSTRACT
Various studies in rodents have shown that nanoparticles are transferred to the breast milk. Under the present study, lactating Wistar rats were repetitively gavaged 5, 25, and 50 mg/kg bw of zinc oxide nanoparticles (ZnO-NPs) and 50 mg kg-1 bw of bulk zinc oxide (bZnO) for 19 days after parturition. The results showed that ZnO-NPs were absorbed in the small intestine of dams and distributed to the liver. Furthermore, ZnO-NPs were distributed to the intestine and liver of rat pups through dam's milk. No significant change in body weight was observed in the dams treated with ZnO-NPs or bZnO and their offsprings as compared to the control group. The spleen weight significantly increased in the rat dams treated with 50 mg kg-1 of ZnO-NPs. ZnO-NPs were mostly excreted through feces. The levels of liver cytochrome P450 reductase and serum total antioxidant capacity significantly decreased in the rat dams treated with ZnO-NPs (50 mg kg-1) and their offsprings. The levels of serum cytokines (tumor necrosis factor-alpha and interleukin-1 beta) and liver injury marker enzymes (alanine aminotransferase and aspartate aminotransferase) significantly increased in the rat dams treated with ZnO-NPs (25 and 50 mg kg-1) and their offsprings. The level of immunoglobulin A secretion in the intestinal fluid of rat dams and their offsprings is significantly increased by increasing the dose of ZnO-NPs. Histopathology of intestine and liver of offsprings whose rat dams were treated with ZnO-NPs (50 mg kg-1) showed gross pathological changes. These results provide information for the safety evaluation of ZnO-NPs use during lactation. In conclusion, a dose-dependent postnatal transfer of ZnO-NPs is hazardous to the breastfed offsprings.