Merck
  • Healthcare resource utilization and direct medical costs associated with index and recurrent Clostridioides difficile infection: a real-world data analysis.
Healthcare resource utilization and direct medical costs associated with index and recurrent Clostridioides difficile infection: a real-world data analysis.
Journal of medical economics (2020-01-31)
Paul Feuerstadt, Laura Stong, David N Dahdal, Naomi Sacks, Kathleen Lang, Winnie W Nelson
ABSTRACT
Aims: This study aimed to evaluate all-cause economic outcomes, healthcare resource utilization (HRU), and costs in patients with Clostridioides difficile infection (CDI) and recurrent CDI (rCDI) using commercial claims from a large database representing various healthcare settings.Materials and methods: A retrospective analysis of commercial claims data from the IQVIA PharMetrics Plus database was conducted for patients aged 18-64 years with CDI episodes requiring inpatient stay with CDI diagnosis code or an outpatient medical claim for CDI plus a CDI treatment. Index CDI episodes occurred between 1 January 2010 and 30 June 2017, including only those where patients were observable 6 months before and 12 months after the index episode. Each CDI episode was followed by a 14-d claim-free period. rCDI was defined as another CDI episode within an 8-week window following the claim-free period. HRU, all-cause direct medical costs and time to rCDI were calculated over 12 months and stratified by number of rCDI episodes.Results: A total of 46,571 patients with index CDI were included. Mean time from one CDI episode to the next was approximately 1 month. In the 12-month follow-up period, those with no recurrence had 1.4 inpatient visits per person and those with 3 or more recurrences had 5.8. Most patients with 3 or more recurrences had 2 or more hospital admissions. The mean annual, total all-cause direct medical costs per patient were $71,980 for those with no recurrence and $207,733 for those with 3 or more recurrences.Limitations: The study included individuals 18-64 years only. A stringent definition of rCDI was used, which may have underestimated the incidence of rCDI.Conclusions: CDI and rCDI are associated with substantial healthcare resource utilization and direct medical costs. Timing of recurrences can be predictable, providing a window of opportunity for interventions. Prevention of multiple rCDI appears essential to reduce healthcare costs.