Merck
  • Thermal responsive poly(N-isopropylacrylamide) grafted chicken feather keratin prepared via surface initiated aqueous Cu(0)-mediated RDRP: Synthesis and properties.
Thermal responsive poly(N-isopropylacrylamide) grafted chicken feather keratin prepared via surface initiated aqueous Cu(0)-mediated RDRP: Synthesis and properties.
International journal of biological macromolecules (2020-02-29)
Sikai Chen, Naruhito Hori, Mikio Kajiyama, Akio Takemura
ABSTRACT
Poultry chicken feather keratin was extracted and then modified for the fabrication of keratin-graft-PNIPAM copolymers. The keratin was well extracted from feather fiber and powdered. Subsequently, it underwent the surficial functionalization process with initiator groups. After the study conducted full disproportionation of Cu(I)Br/Me6Tren into Cu(0) and Cu(II)Br2 in the solvent, surface initiated aqueous Cu(0)-mediated reversible-deactivation radical polymerization (RDRP) of N-isopropylacrylamide (NIPAM) was performed in a methanol/water mixture solvent. The reaction was performed rapidly and efficiently, during which over 100% graft rate was achieved at 60 min. After 6 h reaction, 200% graft rate could be achieved. High graft rate (up to 287%) was achieved, and graft rate could be regulated by controlling the reaction time and the addition of monomer. The fabricated keratin-g-PNIPAM exhibited a rough surface. As revealed from the results of thermal analysis, the thermal stability of keratin-g-PNIPAM was enhanced noticeably compared with the original keratin. Besides, grafted PNIPAM chains exhibited a higher glass transition temperature. The grafted keratin particles displayed enhanced hydrophilicity. Keratin-g-PNIPAMs exhibit a lower LCST comparing to homopolymer and the flocculation in hot water behavior could be controlled by regulating graft rate.