Merck
  • Intestinal restriction of Salmonella Typhimurium requires caspase-1 and caspase-11 epithelial intrinsic inflammasomes.
Intestinal restriction of Salmonella Typhimurium requires caspase-1 and caspase-11 epithelial intrinsic inflammasomes.
PLoS pathogens (2020-04-14)
Shauna M Crowley, Xiao Han, Joannie M Allaire, Martin Stahl, Isabella Rauch, Leigh A Knodler, Bruce A Vallance
ABSTRACT
We investigated the role of the inflammasome effector caspases-1 and -11 during Salmonella enterica serovar Typhimurium infection of murine intestinal epithelial cells (IECs). Salmonella burdens were significantly greater in the intestines of caspase-1/11 deficient (Casp1/11-/-), Casp1-/- and Casp11-/- mice, as compared to wildtype mice. To determine if this reflected IEC-intrinsic inflammasomes, enteroid monolayers were derived and infected with Salmonella. Casp11-/- and wildtype monolayers responded similarly, whereas Casp1-/- and Casp1/11-/- monolayers carried significantly increased intracellular burdens, concomitant with marked decreases in IEC shedding and death. Pretreatment with IFN-γ to mimic inflammation increased caspase-11 levels and IEC death, and reduced Salmonella burdens in Casp1-/- monolayers, while high intracellular burdens and limited cell shedding persisted in Casp1/11-/- monolayers. Thus caspase-1 regulates inflammasome responses in IECs at baseline, while proinflammatory activation of IECs reveals a compensatory role for caspase-11. These results demonstrate the importance of IEC-intrinsic canonical and non-canonical inflammasomes in host defense against Salmonella.