Merck
  • Small Nucleolar RNAs Determine Resistance to Doxorubicin in Human Osteosarcoma.
Small Nucleolar RNAs Determine Resistance to Doxorubicin in Human Osteosarcoma.
International journal of molecular sciences (2020-07-01)
Martina Godel, Deborah Morena, Preeta Ananthanarayanan, Ilaria Buondonno, Giulio Ferrero, Claudia M Hattinger, Federica Di Nicolantonio, Massimo Serra, Riccardo Taulli, Francesca Cordero, Chiara Riganti, Joanna Kopecka
ABSTRACT
Doxorubicin (Dox) is one of the most important first-line drugs used in osteosarcoma therapy. Multiple and not fully clarified mechanisms, however, determine resistance to Dox. With the aim of identifying new markers associated with Dox-resistance, we found a global up-regulation of small nucleolar RNAs (snoRNAs) in human Dox-resistant osteosarcoma cells. We investigated if and how snoRNAs are linked to resistance. After RT-PCR validation of snoRNAs up-regulated in osteosarcoma cells with different degrees of resistance to Dox, we overexpressed them in Dox-sensitive cells. We then evaluated Dox cytotoxicity and changes in genes relevant for osteosarcoma pathogenesis by PCR arrays. SNORD3A, SNORA13 and SNORA28 reduced Dox-cytotoxicity when over-expressed in Dox-sensitive cells. In these cells, GADD45A and MYC were up-regulated, TOP2A was down-regulated. The same profile was detected in cells with acquired resistance to Dox. GADD45A/MYC-silencing and TOP2A-over-expression counteracted the resistance to Dox induced by snoRNAs. We reported for the first time that snoRNAs induce resistance to Dox in human osteosarcoma, by modulating the expression of genes involved in DNA damaging sensing, DNA repair, ribosome biogenesis, and proliferation. Targeting snoRNAs or down-stream genes may open new treatment perspectives in chemoresistant osteosarcomas.