Merck
  • Mycotoxin Determination in Animal Feed: An LC-FLD Method for Simultaneous Quantification of Aflatoxins, Ochratoxins and Zearelanone in This Matrix.
Mycotoxin Determination in Animal Feed: An LC-FLD Method for Simultaneous Quantification of Aflatoxins, Ochratoxins and Zearelanone in This Matrix.
Toxins (2020-06-11)
Borja Muñoz-Solano, Elena González-Peñas
ABSTRACT
Mycotoxins are toxic compounds for humans and animals that are produced by fungi. Mycotoxin contamination in feed is a global safety concern and effective control of these compounds in this matrix is needed. This study proposes a simple, cost-effective analytical method based on liquid chromatography coupled with a fluorescence detector, which is suitable for the routine monitoring of some of the most important mycotoxins in feed: aflatoxins (G2, G1, B2, and B1), zearalenone, and ochratoxins A and B. Mycotoxin extraction, chromatographic separation and quantification are carried out simultaneously for all mycotoxins. The extraction procedure is performed using acetonitrile, water and orthophosphoric acid (80:19:1). Purification of the extract is carried out using an OASIS PRIME HLB solid-phase extraction cartridge followed by a dispersive liquid-liquid microextraction procedure. Aflatoxins G1 and B1 are derivatized post-column (photochemical reactor at 254 nm) to increase their signal. The method has been validated in feed for pigs, cows, sheep, and poultry with very satisfactory results. The detection limits are 2 μg/kg for aflatoxins B1 and G1, 0.64 μg/kg for aflatoxins B2 and G2, 42 μg/kg for zearalenone, and 5 μg/kg for ochratoxins A and B. These values are low enough to allow for monitoring of these mycotoxins in feed. Global recovery values were between 73.6% and 88.0% for all toxins with a relative standard deviation (RSD) % < 7%. This methodology will facilitate laboratory control and analysis of mycotoxins in feed.