Merck
  • piRNA-independent function of PIWIL1 as a co-activator for anaphase promoting complex/cyclosome to drive pancreatic cancer metastasis.
piRNA-independent function of PIWIL1 as a co-activator for anaphase promoting complex/cyclosome to drive pancreatic cancer metastasis.
Nature cell biology (2020-03-24)
Feng Li, Peng Yuan, Ming Rao, Chun-Hui Jin, Wei Tang, Ye-Fei Rong, Yun-Ping Hu, Fengjuan Zhang, Tao Wei, Qi Yin, Tingbo Liang, Ligang Wu, Jinsong Li, Dangsheng Li, Yingbin Liu, Wenhui Lou, Shuang Zhao, Mo-Fang Liu
ABSTRACT
Piwi proteins are normally restricted in germ cells to suppress transposons through associations with Piwi-interacting RNAs (piRNAs), but they are also frequently activated in many types of human cancers. A great puzzle is the lack of significant induction of corresponding piRNAs in cancer cells, as we document here in human pancreatic ductal adenocarcinomas (PDACs), which implies that such germline-specific proteins are somehow hijacked to promote tumorigenesis through a different mode of action. Here, we show that in the absence of piRNAs, human PIWIL1 in PDAC functions as an oncoprotein by activating the anaphase promoting complex/cyclosome (APC/C) E3 complex, which then targets a critical cell adhesion-related protein, Pinin, to enhance PDAC metastasis. This is in contrast to piRNA-dependent PIWIL1 ubiquitination and removal by APC/C during late spermiogenesis. These findings unveil a piRNA-dependent mechanism to switch PIWIL1 from a substrate in spermatids to a co-activator of APC/C in human cancer cells.