Merck
  • Baseline for plastic and hydrocarbon pollution of rivers, reefs, and sediment on beaches in Veracruz State, México, and a proposal for bioremediation.
Baseline for plastic and hydrocarbon pollution of rivers, reefs, and sediment on beaches in Veracruz State, México, and a proposal for bioremediation.
Environmental science and pollution research international (2020-04-26)
Leticia Narciso-Ortiz, Alejandro Coreño-Alonso, Diana Mendoza-Olivares, Carlos Alexander Lucho-Constantino, Manuel Alejandro Lizardi-Jiménez
ABSTRACT
Plastic and hydrocarbon pollution in aquatic ecosystems is a worldwide reality and serious concern today. Plastic debris presents a threat to ecosystems and organisms. Hydrocarbons are also considered priority pollutants. The hydrophobicity of the polymer in combination with the high surface area causes plastics to act as a vector for organic contaminants such as hydrocarbons. The first aim of this work was to evaluate the presence of plastic and hydrocarbon pollution in water from two reefs and two rivers and to identify plastic in six sediment beaches in Veracruz State, Mexico. In addition, the second aim was to analyse the ability of a bacterial consortium to biodegrade hydrocarbons in an airlift bioreactor and to identify degrading bacterial strains of polyethylene terephthalate (PET). Microplastics (100 nm-5 mm) were found in four water samples. Fragments of plastic collected from the reefs ranged in size from 0.716 to 32 μm and in rivers from 0.833 to 784 μm. On the sediment beaches, macroplastics of sizes 2-10 cm were detected. A number of hydrocarbons were also detected in the water samples of both reefs and one river, including n-octane, n-nonane, phenanthrene, n-eicosane, n-dotriacontane, n-hexatriacontane, n-triacontane, and n-tetratriacontane. As a biotechnological alternative for remediation of hydrocarbons and plastics, we attempted to produce a collection of native microorganisms able to degrade them. This work shows results from the bioprospection of a bacterial consortium (Xanthomonas, Acinetobacter bouvetii, Shewanella, and Aquamicrobium lusatiense) for hydrocarbon biodegradation in an airlift bioreactor. The tested consortium was able to successfully degrade the maximum diesel concentration (20 g L-1) tested for 10 days. Also, the first visual evidence of PET degradation by an isolated forest-native bacterial strain showed that Bacillus muralis is the most efficient degrader.