Merck
  • H-Ras induces Nrf2-Pin1 interaction: Implications for breast cancer progression.
H-Ras induces Nrf2-Pin1 interaction: Implications for breast cancer progression.
Toxicology and applied pharmacology (2020-07-06)
Soma Saeidi, Su-Jung Kim, Hyeong-Jun Han, Seong Hoon Kim, Jie Zheng, Han-Byoel Lee, Wonshik Han, Dong-Young Noh, Hye-Kyung Na, Young-Joon Surh
ABSTRACT
Aberrant activation of H-Ras is often associated with tumor aggressiveness in breast cancer. Peptidyl-prolyl cis-trans isomerase NIMA-interacting 1  (Pin1) is a unique enzyme that interacts with phosphorylated serine or threonine of a target protein and isomerizes the adjacent proline residue. Pin1 is prevalently overexpressed in human cancers, and its overexpression correlates with poor prognosis. Nuclear factor E2-related factor 2 (Nrf2) is a master regulator of cellular redox homeostasis. The sustained activation/accumulation of Nrf2 has been observed in many different types of human malignancies, conferring an advantage for growth and survival of cancer cells. The activated form of H-Ras (GTP-H-Ras) is highly overexpressed in human breast cancer tissues. In our present study, silencing of H-Ras decreased the invasiveness of MDA-MB-231 human breast cancer cells and abrogated the interaction between Pin1 and Nrf2 in these cells. Pin1 knockdown blocked the accumulation of Nrf2, thereby suppressing proliferation and clonogenicity of MCF10A-Ras human mammary epithelial cells. We found that Pin1 binds to Nrf2 which stabilizes this transcription factor by hampering proteasomal degradation. In conclusion, H-Ras activation in cooperation with the Pin1-Nrf2 complex represents a novel mechanism underlying breast cancer progression and constitutive activation of Nrf2 and can be exploited as a therapeutic target.