Merck
  • Nitrogen limitation reveals large reserves in metabolic and translational capacities of yeast.
Nitrogen limitation reveals large reserves in metabolic and translational capacities of yeast.
Nature communications (2020-04-22)
Rosemary Yu, Kate Campbell, Rui Pereira, Johan Björkeroth, Qi Qi, Egor Vorontsov, Carina Sihlbom, Jens Nielsen
ABSTRACT
Cells maintain reserves in their metabolic and translational capacities as a strategy to quickly respond to changing environments. Here we quantify these reserves by stepwise reducing nitrogen availability in yeast steady-state chemostat cultures, imposing severe restrictions on total cellular protein and transcript content. Combining multi-omics analysis with metabolic modeling, we find that seven metabolic superpathways maintain >50% metabolic capacity in reserve, with glucose metabolism maintaining >80% reserve capacity. Cells maintain >50% reserve in translational capacity for 2490 out of 3361 expressed genes (74%), with a disproportionately large reserve dedicated to translating metabolic proteins. Finally, ribosome reserves contain up to 30% sub-stoichiometric ribosomal proteins, with activation of reserve translational capacity associated with selective upregulation of 17 ribosomal proteins. Together, our dataset provides a quantitative link between yeast physiology and cellular economics, which could be leveraged in future cell engineering through targeted proteome streamlining.