Merck
  • SRF Potentiates Colon Cancer Metastasis and Progression in a microRNA-214/PTK6-Dependent Manner.
SRF Potentiates Colon Cancer Metastasis and Progression in a microRNA-214/PTK6-Dependent Manner.
Cancer management and research (2020-08-18)
Tao Li, Yingchun Wan, Ziyuan Su, Jiayu Li, Minna Han, Changyu Zhou
ABSTRACT
Serum response factor (SRF), a sequence-specific transcription factor, is closely related to metastasis of gastric cancer, a digestive tract cancer. Herein, we probed the effect of SRF on metastasis and progression of colon cancer (CC), another digestive tract disorder, and the detailed mechanism. Microarray analysis was conducted on tumor and adjacent tissues to filter differentially expressed miRNA, followed by RT-qPCR validation in CC cell lines. The transcription factor and the target gene of microRNA-214 (miR-214) were predicted, and their binding relationships were tested by luciferase reporter assays and ChIP assays. Subsequently, SRF and protein tyrosine kinase 6 (PTK6) expression in CC patients and cells was evaluated by RT-qPCR, while JAK2 and STAT3 expression in cells by Western blot analysis. To further explore functions of miR-214, PTK6 and SRF on CC, CC cells were delivered with si-PTK6, miR-214 mimic and/or SRF overexpression. miR-214 expressed poorly in CC tissues and cell lines, which related to advanced TNM staging and survival. miR-214 mimic inhibited proliferation, migration, invasion, xenograft tumor growth and metastasis of CC cells. SRF, overexpressed in CC samples and cells, suppressed the transcription of miR-214. Meanwhile, SRF upregulation counteracted the inhibitory role of miR-214 mimic in CC cell growth. miR-214 negatively regulated PTK6 expression to impair the JAK2/STAT3 pathway activation, thereby halting CC cell proliferation, migration, invasion, xenograft tumor growth and metastasis. Altogether, miR-214 may perform as a tumor suppressor in CC, and the SRF/miR-214/PTK6/JAK2/STAT3 axis could be applied as a biomarker and potential therapeutic target.