Merck
  • Profiling of naïve and primed human pluripotent stem cells reveals state-associated miRNAs.
Profiling of naïve and primed human pluripotent stem cells reveals state-associated miRNAs.
Scientific reports (2020-07-01)
Benjamin T Dodsworth, Klas Hatje, Maria Rostovskaya, Rowan Flynn, Claas A Meyer, Sally A Cowley
ABSTRACT
Naïve human pluripotent stem cells (hPSC) resemble the embryonic epiblast at an earlier time-point in development than conventional, 'primed' hPSC. We present a comprehensive miRNA profiling of naïve-to-primed transition in hPSC, a process recapitulating aspects of early in vivo embryogenesis. We identify miR-143-3p and miR-22-3p as markers of the naïve state and miR-363-5p, several members of the miR-17 family, miR-302 family as primed markers. We uncover that miR-371-373 are highly expressed in naïve hPSC. MiR-371-373 are the human homologs of the mouse miR-290 family, which are the most highly expressed miRNAs in naïve mouse PSC. This aligns with the consensus that naïve hPSC resemble mouse naive PSC, showing that the absence of miR-371-373 in conventional hPSC is due to cell state rather than a species difference.