Merck
  • Binding properties of sodium glucose co-transporter-2 inhibitor empagliflozin to human serum albumin: spectroscopic methods and computer simulations.
Binding properties of sodium glucose co-transporter-2 inhibitor empagliflozin to human serum albumin: spectroscopic methods and computer simulations.
Journal of biomolecular structure & dynamics (2019-08-06)
Wenjing Wang, Na Gan, Qiaomei Sun, Di Wu, Ludan Zhao, Zili Suo, Peixiao Tang, Hui Li
ABSTRACT
Empagliflozin is an oral sodium glucose co-transporter-2 inhibitor for type 2 diabetes mellitus. The interaction between empagliflozin and human serum albumin (HSA) was investigated experimentally and theoretically. Fluorescence quenching and time-resolved fluorescence spectroscopy indicated that the quenching mechanism of empagliflozin and HSA was dynamic and that the effective binding constant at body temperature was 3.495 × 103 M-1. Thermodynamic parameters showed that hydrophobic forces were the major binding force in the interaction between empagliflozin and HSA. Circular dichroism, Fourier transform infrared, and 3 D fluorescence spectroscopy revealed that empagliflozin showed a slight change in secondary structure without changing the basic carbon framework of HSA. Site marker displacement experiments revealed that empagliflozin bound to site I of HSA, which was supported by molecular docking. Molecular dynamic simulations indicated that empagliflozin could bind to HSA stably. This study provided insights into the binding mechanism between empagliflozin and HSA.Communicated by Ramaswamy H. Sarma.