Merck
  • Inhibition of lipoprotein-associated phospholipase A2 reduces complex coronary atherosclerotic plaque development.
Inhibition of lipoprotein-associated phospholipase A2 reduces complex coronary atherosclerotic plaque development.
Nature medicine (2008-09-23)
Robert L Wilensky, Yi Shi, Emile R Mohler, Damir Hamamdzic, Mark E Burgert, Jun Li, Anthony Postle, Robert S Fenning, James G Bollinger, Bryan E Hoffman, Daniel J Pelchovitz, Jisheng Yang, Rosanna C Mirabile, Christine L Webb, LeFeng Zhang, Ping Zhang, Michael H Gelb, Max C Walker, Andrew Zalewski, Colin H Macphee
ABSTRACT
Increased lipoprotein-associated phospholipase A(2) (Lp-PLA(2)) activity is associated with increased risk of cardiac events, but it is not known whether Lp-PLA(2) is a causative agent. Here we show that selective inhibition of Lp-PLA(2) with darapladib reduced development of advanced coronary atherosclerosis in diabetic and hypercholesterolemic swine. Darapladib markedly inhibited plasma and lesion Lp-PLA(2) activity and reduced lesion lysophosphatidylcholine content. Analysis of coronary gene expression showed that darapladib exerted a general anti-inflammatory action, substantially reducing the expression of 24 genes associated with macrophage and T lymphocyte functioning. Darapladib treatment resulted in a considerable decrease in plaque area and, notably, a markedly reduced necrotic core area and reduced medial destruction, resulting in fewer lesions with an unstable phenotype. These data show that selective inhibition of Lp-PLA(2) inhibits progression to advanced coronary atherosclerotic lesions and confirms a crucial role of vascular inflammation independent from hypercholesterolemia in the development of lesions implicated in the pathogenesis of myocardial infarction and stroke.