Merck
  • Identification of ERGIC-53 as an intracellular transport receptor of alpha1-antitrypsin.
Identification of ERGIC-53 as an intracellular transport receptor of alpha1-antitrypsin.
The Journal of cell biology (2008-02-20)
Beat Nyfeler, Veronika Reiterer, Markus W Wendeler, Eduard Stefan, Bin Zhang, Stephen W Michnick, Hans-Peter Hauri
ABSTRACT
Secretory proteins are exported from the endoplasmic reticulum (ER) by bulk flow and/or receptor-mediated transport. Our understanding of this process is limited because of the low number of identified transport receptors and cognate cargo proteins. In mammalian cells, the lectin ER Golgi intermediate compartment 53-kD protein (ERGIC-53) represents the best characterized cargo receptor. It assists ER export of a subset of glycoproteins including coagulation factors V and VIII and cathepsin C and Z. Here, we report a novel screening strategy to identify protein interactions in the lumen of the secretory pathway using a yellow fluorescent protein-based protein fragment complementation assay. By screening a human liver complementary DNA library, we identify alpha1-antitrypsin (alpha1-AT) as previously unrecognized cargo of ERGIC-53 and show that cargo capture is carbohydrate- and conformation-dependent. ERGIC-53 knockdown and knockout cells display a specific secretion defect of alpha1-AT that is corrected by reintroducing ERGIC-53. The results reveal ERGIC-53 to be an intracellular transport receptor of alpha1-AT and provide direct evidence for active receptor-mediated ER export of a soluble secretory protein in higher eukaryotes.