Merck
  • Stability of emulsions stabilised by two physiological surfactants: L-alpha-phosphatidylcholine and sodium taurocholate.
Stability of emulsions stabilised by two physiological surfactants: L-alpha-phosphatidylcholine and sodium taurocholate.
Biophysical chemistry (1995-05-01)
A J Fillery-Travis, L H Foster, M M Robins
ABSTRACT
The emulsion phase formed within the stomach and duodenum during digestion of a fatty meal has been modelled using two physiological surfactants, the phospholipid L-alpha-phosphatidylcholine (PC) and the bile salt sodium taurocholate (NaT). Upon dilution of the phospholipid stabilised emulsions with a solution of NaT the bile salt became incorporated into the oil/water interface imparting a negative charge to the droplet surface. The magnitude of the droplet microelectrophoretic mobility for the mixed PC and NaT system was 47% of that found for emulsion droplets stabilised by NaT alone. But the electrostatic repulsion between droplets was not sufficient to account for the observed improvement in emulsion stability to coalescence. It is suggested that a residual liquid crystalline phospholipid interface is present imparting a significant steric component to the stabilisation of the emulsions droplets.