Merck
  • Crystallization of a 2:2 complex of granulocyte-colony stimulating factor (GCSF) with the ligand-binding region of the GCSF receptor.
Crystallization of a 2:2 complex of granulocyte-colony stimulating factor (GCSF) with the ligand-binding region of the GCSF receptor.
Acta crystallographica. Section F, Structural biology and crystallization communications (2006-03-03)
Eijiro Honjo, Taro Tamada, Yoshitake Maeda, Takumi Koshiba, Yasuko Matsukura, Tomoyuki Okamoto, Matsujiro Ishibashi, Masao Tokunaga, Ryota Kuroki
ABSTRACT
The granulocyte-colony stimulating factor (GCSF) receptor receives signals for regulating the maturation, proliferation and differentiation of the precursor cells of neutrophilic granulocytes. The signalling complex composed of two GCSFs (GCSF, 19 kDa) and two GCSF receptors (GCSFR, 34 kDa) consisting of an Ig-like domain and a cytokine-receptor homologous (CRH) domain was crystallized. A crystal of the complex was grown in 1.0 M sodium formate and 0.1 M sodium acetate pH 4.6 and belongs to space group P4(1)2(1)2 (or its enantiomorph P4(3)2(1)2), with unit-cell parameters a = b = 110.1, c = 331.8 A. Unfortunately, this crystal form did not diffract beyond 5 A resolution. Since the heterogeneity of GCSF receptor appeared to prevent the growth of good-quality crystals, the GCSF receptor was fractionated by anion-exchange chromatography. Crystals of the GCSF-fractionated GCSF receptor complex were grown as a new crystal form in 0.2 M ammonium phosphate. This new crystal form diffracted to beyond 3.0 A resolution and belonged to space group P3(1)21 (or its enantiomorph P3(2)21), with unit-cell parameters a = b = 134.8, c = 105.7 A.