Merck
  • Effects of adenosine A1-receptor activation on torsade de pointes in rabbits.
Effects of adenosine A1-receptor activation on torsade de pointes in rabbits.
Cardiovascular drugs and therapy (1999-11-05)
H L Tan, C J Hou, R J Sung
ABSTRACT
We tested whether the adenosine A1 receptor agonist, R-PIA, suppressed torsade de pointes (TdP) induced by the delayed rectifier potassium channel blocker clofilium. Furthermore, we studied the underlying mechanism: beta-adrenergic antagonism or ATP-sensitive K+ channel (IK-ATP) opening. In anesthetized rabbits, TdP was induced by simultaneous infusion of clofilium and the alpha1-adrenoceptor agonist methoxamine. Four groups were studied: (1) saline infusion after TdP induction; (2) R-PIA (1.3 mg/kg) infusion; (3) R-PIA infusion after propranolol (2 micromol/kg) pretreatment; (4) R-PIA infusion after glibenclamide (10 micromol/kg) pretreatment. TdP suppression rate was 0% in group 1, 78% in group 2 (p<0.01 vs. group 1), 67% in group 3 (p<0.05 vs. group 1, p = NS vs. group 2), 33% in group 4 (p = NS vs. group 1, p = 0.08 vs. group 2). TdP induction coincided with increased QT/QTc duration and QT dispersion. TdP suppression coincided with reduced QT dispersion, but further QT/QTc lengthening. R-PIA suppressed TdP, not by beta-adrenergic antagonism, but mostly by IK-ATP opening. QT dispersion correlated better with TdP induction/suppression than QT/QTc duration.