- Fluorescent phallotoxin, a tool for the visualization of cellular actin.
Fluorescent phallotoxin, a tool for the visualization of cellular actin.
Proceedings of the National Academy of Sciences of the United States of America (1979-09-01)
E Wulf, A Deboben, F A Bautz, H Faulstich, T Wieland
PMID291981
ABSTRACT
A fluorescent derivative of phalloidin has been synthesized possessing high affinity to filamentous actin. This compound was used for visualization of actin-containing structures in eukaryotic nonmuscle cells. Due to its low molecular weight (1250), fixation for formaldehyde was sufficient to render the membrane permeable for the labeled peptide. Bundles of microfilaments are the predominant pattern in the flat rat kangaroo PtK1 cells, whereas a net of concentric fibers characterizes the more spherical bovine kidney MDBK cells. Specificity of staining was confirmed by competition experiments with unlabeled phalloidin.
Sigma-Aldrich
Phalloidin, Fluorescein Isothiocyanate Labeled, sequence Amanita phalloides(synthetic: peptide sequence)