Merck
  • Characterization and relative quantification of phospholipids based on methylation and stable isotopic labeling.
Characterization and relative quantification of phospholipids based on methylation and stable isotopic labeling.
Journal of lipid research (2016-01-07)
Tanxi Cai, Qingbo Shu, Peibin Liu, Lili Niu, Xiaojing Guo, Xiang Ding, Peng Xue, Zhensheng Xie, Jifeng Wang, Nali Zhu, Peng Wu, Lili Niu, Fuquan Yang
ABSTRACT
Phospholipids (PLs), one of the lipid categories, are not only the primary building blocks of cellular membranes, but also can be split to produce products that function as second messengers in signal transduction and play a pivotal role in numerous cellular processes, including cell growth, survival, and motility. Here, we present an integrated novel method that combines a fast and robust TMS-diazomethane-based phosphate derivatization and isotopic labeling strategy, which enables simultaneous profiling and relative quantification of PLs from biological samples. Our results showed that phosphate methylation allows fast and sensitive identification of the six major PL classes, including their lysophospholipid counterparts, under positive ionization mode. The isotopic labeling of endogenous PLs was achieved by deuterated diazomethane, which was generated through acid-catalyzed hydrogen/deuterium (H/D) exchange and methanolysis of TMS-diazomethane during the process of phosphate derivatization. The measured H/D ratios of unlabeled and labeled PLs, which were mixed in known proportions, indicated that the isotopic labeling strategy is capable of providing relative quantitation with adequate accuracy, reproducibility, and a coefficient of variation of 9.1%, on average. This novel method offers unique advantages over existing approaches and presents a powerful tool for research of PL metabolism and signaling.