- Stereoselective synthesis of myo-inositol via ring-closing metathesis: a building block for glycosylphosphatidylinositol (GPI) anchor synthesis.
Stereoselective synthesis of myo-inositol via ring-closing metathesis: a building block for glycosylphosphatidylinositol (GPI) anchor synthesis.
Organic letters (2002-04-13)
Rosemary M Conrad, Michael J Grogan, Carolyn R Bertozzi
PMID11950362
ABSTRACT
Here we report a concise stereoselective synthesis of myo-inositol via ring-closing metathesis. A readily available bis-Weinreb amide of D-tartrate served as a key intermediate. [reaction: see text]