Merck
  • Long non-coding RNA HOTAIR knockdown enhances radiosensitivity through regulating microRNA-93/ATG12 axis in colorectal cancer.
Long non-coding RNA HOTAIR knockdown enhances radiosensitivity through regulating microRNA-93/ATG12 axis in colorectal cancer.
Cell death & disease (2020-03-08)
Yingqiang Liu, Xijuan Chen, Xiling Chen, Junqi Liu, Hao Gu, Ruitai Fan, Hong Ge
ABSTRACT
Colorectal cancer (CRC) is a global healthcare problem. Radioresistance is a huge setback for CRC radiotherapy. In this text, the roles and molecular mechanisms of long non-coding RNA HOTAIR in CRC tumorigenesis and radioresistance were further investigated. ATG12 mRNA, HOTAIR, and microRNA-93 (miR-93) levels were measured by quantitative reverse transcription polymerase chain reaction (RT-qPCR) assay. Protein levels of LC3 I, LC3 II, p62, ATG12, cleaved caspase 3, Bax, and Bcl-2 were detected by western blotting assay in cells and were examined by immunohistochemistry (IHC) assay in tissues. Cell survival fractions, viability, and apoptotic rates were determined by clonogenic survival assay, CCK-8 assay, and flow cytometry analysis, respectively. The relationships of HOTAIR, miR-93, and ATG12 were tested by bioinformatics analysis and luciferase reporter assay. Mouse xenograft tumor models were established to investigate the influence of HOTAIR knockdown on CRC radioresistance in vivo. We found that HOTAIR expression was markedly upregulated in plasma from CRC patients after radiotherapy and CRC cells after irradiation. HOTAIR knockdown, miR-93 overexpression, or ATG12 silencing weakened cell viability, induced cell apoptosis, inhibited cell autophagy, and enhanced cell radiosensitivity in CRC. HOTAIR exerted its functions by downregulating miR-93. Moreover, HOTAIR functioned as a molecular sponge of miR-93 to regulate ATG12 expression. ATG12 protein expression was markedly upregulated and associated with miR-93 and HOTAIR expression in CRC tissues. Furthermore, HOTAIR knockdown enhanced radiosensitivity of CRC xenograft tumors by regulating miR-93/ATG12 axis. In conclusion, HOTAIR knockdown potentiated radiosensitivity through regulating miR-93/ATG12 axis in CRC, further elucidating the roles and molecular basis of HOTAIR in CRC radioresistance.